Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
General-Purpose Open Drip-Proof (ODP) Motors
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX71011-11EA 67832.08 INR InStock
MFLX71011-11
General-Purpose Open Drip-Proof (ODP) Motors
Pumps Motors
Use on pumps, compressors, fans, blowers, and other machinery.
  • Single-phase motors have a resilient base — mount motor horizontally or vertically
Single-phase motors have a removable, rigid base. Both single-phase motors and models 70011-04 and -16 (three-phase motors)
feature automatic overload protection.
Three-phase motors include a rigid base. Models 70011-28 to -92 have a NEMA conduit box.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms