Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Stainless Steel Compression to NPT(M) Adapter Elbows
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
31406-78 31406-75 31406-79 31406-81
MFLX31406-78EA 5661.17 INR InStock
MFLX31406-78 MFLX31406-75 MFLX31406-79 MFLX31406-81
Stainless Steel Compression to NPT(M) Adapter Elbows
Fittings and Connectors
Create a secure connection with compression fittings.
  • Constructed of durable stainless steel (SS)
Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC).
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms