Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Flowline EchoSafe Explosion-Proof Ultrasonic Level transmitter (2-wire)
Supplier: Flowline
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX68873-81EA 305101.45 INR InStock
MFLX68873-81
Flowline EchoSafe Explosion-Proof Ultrasonic Level transmitter (2-wire)
Level Sensors
Explosion-proof ultrasonic level transmitter ideal for hazardous applications.
  • Compact XP enclosure with viewing window and dual conduit ports
  • Narrow 3'' beam diameter enables flexible installation in applications with limited measurement space
  • Rugged corrosion resistant PVDF transducer and process mount for chemical and wastewater
  • Fail-safe intelligence with diagnostic feedback for easy troubleshooting
  • Display indicates air gap or level height distance in inch or centimeter
EchoSafeâ„¢, an explosion proof, two-wire ultrasonic transmitter provides noncontact level measurement up to 10m in hazardous bulk storage, day tank and sump applications, and is well suited for challenging corrosive, slurry or waste media. Media examples include sodium hypochlorite, wastewater and resin. The integral display and push button module enable fast and simple calibration.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms