Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Thermo Scientific Megafuge 16 and 16R Centrifuges
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
17707-41 17707-51
MFLX17707-41EA 0 INR InStock
MFLX17707-41 MFLX17707-51
Thermo Scientific Megafuge 16 and 16R Centrifuges
Centrifuges Benchtop Centrifuges
Designed around you and your applications.
  • AutoLock® III rotor system provides easy rotor change in about 3-seconds!
  • Motorized lid-latch reduces wear and closure time
  • Intuitive display and control for routine and repetitive environments — perfect for clinical protocols, cell culture applications, and microplate processing
These general-purpose centrifuges provide flexibility with an easy change rotor system, and improved safety features. Unique, AutoLock® rotor system makes changes of rotors and buckets easy. With the push of a button, change the rotor — no need for tools. SMARTSpin® rotor management system maximizes safety and separation by optimizing acceleration, braking, and residual load imbalance for any rotor and bucket option.
The compact footprint minimizes the lab space required, while the sleek design provides improved ergonomics and easy clean-up. Motorized lid latch requires minimal effort for closing. Six program keys allow for easier recall of frequently used settings to save time.
Delivery information: Includes power cable, instruction manual, and anti-corrosion oil.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms