ODE ATEX-Rated Solenoid Valves, 316 Stainless Steel
Supplier: ODE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX08610-42EA 140000 INR
MFLX08610-42
ODE ATEX-Rated Solenoid Valves, 316 Stainless Steel
Valves Solenoid Valves
ATEX-rated explosion-proof coils are designed for petrochemical and other industrial applications.
  • Two way normally closed valves offer port sizes for many applications
  • Body material of 316 stainless steel withstands costic media
  • Max operating pressure of 230 psi withstands high-pressure applications
These ODE ATEX-rated solenoid valves are ideal for petrochemical and other industrial applications. Valves are constructed with a 316 stainless steel (SS) body and seat with Viton® seals. Valves feature an EX rating of ATEX directive 94/9 CE Class 2, Div 2 II 2G Ex d II C T6.