Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Economical Submersible Pumps, High-Flow Utility
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX75500-05EA 49043.72 INR InStock
MFLX75500-05 MFLX75500-00
Economical Submersible Pumps, High-Flow Utility
Pumps Centrifugal Pumps
Versatile – operate pumps in either a vertical or horizontal position.
  • Compact and economical
  • Screened inlet helps prevent clogging
High-flow submersible utility pumps for use for water transfer with garden hose adpater or recirculation, water displays, air conditioners, and machine tool coolants. Removable inlet screen allows for water removal 1/8" from floor. Oil-filled for longer motor life. UL listed, CSA certified.
Note: Pumps must be fully submersed when operating and not for use with salt water or flammable solutions.
Delivery information: 115 VAC models include grounded plug.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms