ProtEX-F&I Explosion-Proof Feet & Inches Level Meter
Supplier: Precision Digital
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX30004-92EA 193732.28 INR
MFLX30004-92
ProtEX-F&I Explosion-Proof Feet & Inches Level Meter
Level Sensors
Displays level in feet and inches.
  • 32-point linearization
  • Tank level bar graph
  • Lower display for custom tag, volume, or percent
  • Open collector output for high or low alarm
The ProtEX-F&I PD6801 explosion-proof level meter brings enhanced functionality and readability to hazardous areas in a unique way — it displays level in feet and inches, not a decimal, so it can be read and understood effortlessly. The meter can display in eighths or sixteenths of an inch. Display features a backlight, and can be seen under varied lighting conditions, from wide angles, and from a distance. Program the meter using SafetTouch® thru-glass buttons, eliminating the need to remove the cover.