Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Masterflex® Direct-Reading Variable-Area Flowmeters with Valve, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Masterflex®
MFLX34500-81EA 93573.66 INR InStock
MFLX34500-81 MFLX34500-80 MFLX34500-84 MFLX34500-83
Masterflex® Direct-Reading Variable-Area Flowmeters with Valve, Avantor®
Flow Meters
Durable valve allows for easy control of your media.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Yellow colored backing enhances readability and minimizes eye fatigue
  • Use with various media for laboratory and industrial applications
These variable-area flowmeters are offered with direct reading scales for water, argon, oxygen, carbon dioxide, nitrogen or helium. Multi-turn valve is included, and dual scales display flow rates in both metric and English units. Optional scales can be developed for diverse flow conditions facilitating OEM applications. Flowmeters are configured for in-line 3/4" NPT(F) connections.
Scales can be purchased and interchanged. To change the scales, simply remove two small screws on front plate of meter and remove the plastic cover, and replace scale with desired scale, and replace cover and screws. For more details, contact an Applications Specialist.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms