Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Masterflex® Sanitary Adapters, Animal-Derived Ingredient Free Polypropylene
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX31512-59EA 88300 INR InStock
MFLX31512-59 MFLX31512-15
Masterflex® Sanitary Adapters, Animal-Derived Ingredient Free Polypropylene
Fittings and Connectors
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing.
All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI. Available with 3/8'' or 1/2'' hose barbs and 3/4'' mini or 1-1/2'' standard sanitary clamp connections. Pair with Cole-Parmer platinum-cured silicone sanitary gaskets for a completely animal-derived ingredient-free connection. Pack of 10.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms