You'll be up to speed and pumping in no time at all.
- Integrated pump and drive systems are provided fully assembled, reducing setup time
- Contamination-free pumping—the fluid contacts only the tubing
- Intuitive keypad is easy to use — quickly increase or decrease flow
- Digital display of speed (rpm) for accurate control and repeatability
- Reversible motor permits purging of tubing before and after use
- Pump head accepts multiple tubing sizes to provide a broad flow range
- Unique "rapid-load" pump head allows fast tube loading and minimizes downtime
- Interlock switch powers unit down when pump head is opened for tubing change
- Remote control capability is ideal for automated process applications
Catalyst™ FH100 and FH100X peristaltic pumps are designed specifically for easy repeatability, fluid transfer, metering, and worry-free performance in a variety of life science and laboratory applications. The broad flow range capability of these pumps make them ideal for laboratory to pilot process scale-up requirements. For control flexibility, these pumps feature remote operation of speed, pumping direction, and start/stop.
These pumps are a reliable alternative to piston pumps, gear pumps, and other circulating pumps. Catalyst FH100 variable-speed pumps are self priming and contain no valves or seals — eliminating routine maintenance demands. Fluid contacts only the tubing, providing for contamination-free pumping in high-purity applications.
Delivery information: Includes 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country-specific plug/cord set; please specify ultimate destination when ordering.