Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Catalyst FH-Series Multichannel Peristaltic Pump Cartridges
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX77724-60EA 18500 INR InStock
MFLX77724-60 MFLX77724-65 MFLX77724-70 MFLX77724-75
Catalyst FH-Series Multichannel Peristaltic Pump Cartridges
Pumps Peristaltic Pumps
Use with Catalyst multichannel cartridge pump heads for synchronous, multichannel pumping.
  • Cartridges feature accurate flow rates and adjustable tubing retainers to hold tubing in place
All cartridges have adjustable occlusion which allows for bi-directional flow variation of ±5%. Tighten occlusion for higher pressures, loosen for longer tubing life. Combine the input/output of two cartridges to reduce pulsation.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms