Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Fixing Clamps for Domnick Hunter Air Purification Systems
Supplier: Parker
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX29587-84EA 34026.18 INR InStock
MFLX29587-84
Fixing Clamps for Domnick Hunter Air Purification Systems
Air Purifiers
Optimize performance of your Parker® domnick hunter Air Purification Systems.
  • Suitable for use with with 29587-series Domnick Hunter Air Purification Systems
Fixing/Mounting Clamps join adjacent filters mounted in series. Clamp also functions as a wall-mounting bracket to minimize associated hardware. These clamps are compatible with 29587-series filters.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms