Mini sanitary end cap, 3/4",Natural Kynar PVDF, pack of 10
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX15959-01EA 11500 INR
MFLX15959-01
Mini sanitary end cap, 3/4",Natural Kynar PVDF, pack of 10
Fittings and Connectors
All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow, no particulate build-up and easy cleaning. Made from animal-derived ingredient-free polypropylene and PVDF, these adapters may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI.