Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Sanitary end cap, 1-1/2", Natural Kynar PVDF, pack of 10
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MFLX15959-02EA 11200 INR InStock
MFLX15959-02
Sanitary end cap, 1-1/2", Natural Kynar PVDF, pack of 10
Fittings and Connectors
  • All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow, no particulate build-up and easy cleaning.
  • Made from animal-derived ingredient-free polypropylene and PVDF, these adapters may be sterilized by gamma irradiation.
  • Material meets the requirements for USP Class VI.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms