LinkTech 40AC & 40PP Quick-Disconnect Fitting Insert, Hosebarb without Valve, Panel Mount
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX41304-16EA 25600 INR
MFLX41304-16
LinkTech 40AC & 40PP Quick-Disconnect Fitting Insert, Hosebarb without Valve, Panel Mount
Fittings and Connectors
Easy to disconnect from 1/4" flow line with one hand.
  • Withstands pressures from vacuum to 120 psi (8.3 bar)
  • Interchangeable with other similar industry thumb-latch fittings
Just press the metal thumb-latch to connect and disconnect fitting halves. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized.
Mix and match valved and straight-through (nonvalved) fittings to suit your flow path needs—simply order a body (female half) and an insert (male half). Valved fittings automatically seal the flow path when disconnected.