Corning Centrifuge Tube Caps and Lids
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34534-16 34534-18 34534-20 34534-22
MFLX34534-16EA 0 INR
MFLX34534-16 MFLX34534-18 MFLX34534-20 MFLX34534-22
Corning Centrifuge Tube Caps and Lids
Closures Test/Sample Tube Closures
Choose the cap that fits your application needs.
  • Various liners for multiple uses
Select either disposable phenolic caps with rubber liners, or reusable screw caps with PTFE liners for improved chemical resistance.