Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Volumetric Pipettes, ASTM
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
612-6917EA 4200.17 INR InStock
612-6917 612-6918 612-6919 612-6920 612-6935 612-6936 612-6937 612-6938 612-6939 612-6940 612-6941 612-6942 612-6943 612-6944 612-6945 612-6946 612-6947 612-6948 612-6949
VWR®, Volumetric Pipettes, ASTM
Pipettes Graduated Pipettes
VWR® borosilicate glass pipettes are made from specially selected accurate bore glass tubing. They have precision formed jets, and bevel ground ends to resist chipping. The mouth piece of the pipette is also tooled for easy sucking and cotton plugging.
  • Calibrated at 20 °C
  • Calibrated for delivery (TD, Ex)
  • Generally used for accurate measurement and decanting of liquid
  • Markings in permanent amber stain diffused into the surface of the glass for better visibility and durability
Each pipette is calibrated to deliver (Ex, TD) at 20 °C reference temperature. The calibration also takes into account surface adhesion effects.
A calibration certificate is provided with each Individual certified product.
Certifications: Pipettes are in compliance with ASTM E 969 specification.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms