Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Cabinet Accessories
Supplier: Justrite
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
JUSU24167EA 0 INR InStock
JUSU25950 JUSU29907 JUSU24167 JUSU25933
Cabinet Accessories
Furniture Shelves
Cabinet accessories offer an opportunity to expand storage and add new convenience to existing cabinets or customise new cabinets with additional shelves to meet specific workflow needs.
  • Expand storage
  • Add new convenience
  • Customize new cabinets
Polyethylene trays offer additional protection and have a slightly raised curved edge to contain spills
SpillSlopeâ„¢ galvanised steel shelves hold up to 159 kg (350 lbs.) and adjust on 57 mm increments for versatile storage.
They may also be used in the bottom sill area to catch incidental spills and leaks.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms