Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

BeneSphera™ HCV Rapid Card Test

Supplier: Avantor

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

BeneSphera™
BENP15001481EA 1759.5 INR InStock
BENP15001481
BeneSphera™ HCV Rapid Card Test
Assays Immunological Assays
BeneSphera™ HCV rapid card test is for detection of antibodies to HCV from human whole blood, serum, and plasma.

  • Use of highly purified recombinant HCV antigens for Core, NS3,NS4, NS5
  • Detects all subtypes of HCV
  • Better background clearance and line intensity
  • Sensitive to detect low antibody titre
  • Sensitivity: 99,1%
  • Specificity: 99,6%

BeneSphera™ HCV Rapid Card test is a rapid test to qualitatively detect the presence of antibody to HCV in a human serum or plasma. The BeneSphera™ HCV Rapid Card test has been compared with a leading commercial HCV ELISA test. The correction between these two systems is >99%.

Certifications: National Institute of Biologicals (MoHFW, Govt. of India).

Order Now
 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms