Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Sterile Cleanroom Tapes
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
114-9461EA 0 INR InStock
114-9461 114-9463 VWRA114-9462
VWR® Sterile Cleanroom Tapes
Tapes
VWR® offers a cost-effective and labor-saving range of sterile cleanroom tapes for critical areas. These tapes are compliant and formulated per pharmacopea, ISO, and other international standards specifications.
  • Ideal for use in pharma, biopharma, isolators, and critical cleanrooms
  • Cleanroom tapes are gamma irradiated 10⁻⁶ SAL level
  • Not autoclavable
  • Shelf life: 24 months
  • Triple-wrap packaging
  • Customised packaging is available
VWR® sterile cleanroom tape holds up to wipe down with IPA, bleach, and phenols. Integrity of printed barrel will be compromised with thinners and strong solvents.
Tape rolls are individually bagged, four per double bag, 12 per third bag. Lot numbers and IR dot on each outer bag and on each case.
Minimum irradiation dose 25 kGy.
Certifications: Batchwise CoA and MSDS available as recommended in USP guidelines.
Delivery information: Certificate of Irradiation and Certificate of Conformance shipped with each order.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms