Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Select Bluing Reagent
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRMNPSL403EA 15859 INR InStock
VWRMNPSL403
Select Bluing Reagent
Stains Microscopy and staining reagents
A buffered alkaline rinse with a mild pH (8,0).
  • Ensures optimal staining and proper cellular hue staining
  • Premixed and ready- to-use
  • Tinted blue for easy identification
Part of the StatLab Select stain line.
Manufactured in USA.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms