VWR® Stirring Rods
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
VWRU89026-290EA 17617 INR
VWRU89026-290
VWR® Stirring Rods
Stirrers Stirring Rods
Available with a steel core or in solid PTFE, these rods are chemically resistant, inert, and non-abrasive.
  • Constructed from solid PTFE or with a steel core
  • Chemically resistant
  • Inert
  • Non-abrasive
Steel core rods have a rounded end, can be bent into a permanent shape, and are ideal when extra rigidity is required. Solid PTFE rods have a tapered end. Usable to 280 °C (536 °F).