Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Jaw Style Tubing Clamp, Acetal
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU70690-116EA 3620 INR InStock
VWRU70690-116
VWR® Jaw Style Tubing Clamp, Acetal
Clamps Tubing and Sanitary Clamps
Tube clamp is molded of strong, durable plastic with serrated jaws 270 mm (1¹⁄₁₆") long.
  • Designed for one-handed operation
  • Strong, durable plastic construction
  • Serrated jaws
  • Autoclavable
Attach at any point on tubing without disconnecting tubing
When not in use, the circular opening (approximately 60 mm (¼") diameter) at hinge end allows clamp to remain attached to tubing having 6 to 100 mm (¼ to ⅜") OD with little to no restriction in flow. Overall length is 590 mm (2⁵⁄₁₆"). Autoclavable.
Designed to be operated with one hand, the clamp locks closed and opens when the locking tab is released. For use on tubing up to 130 cm (½") O.D. and wall thickness of 0,82 mm (1/32") or thinner.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms