3,3',5,5'-Tetramethylbenzidine (TMB) ≥98%, BioChemica®

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A3840.0001EA 0 INR
A3840.0001 A3840.0025
3,3',5,5'-Tetramethylbenzidine (TMB) ≥98%, BioChemica®
3,3',5,5'-Tetramethylbenzidine (TMB)
Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 166 to 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00007748
CAS Number: 54827-17-7
EINECS: 259-364-6
UN: 0000

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR