Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Hematology Analyser, BeneSphera™ H33S - RFID
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MIND3008D-PA00009EA 396000 INR InStock
MIND3008D-PA00009
Hematology Analyser, BeneSphera™ H33S - RFID
Clinical Diagnostic Supplies Clinical Analysers
The BeneSphera™ H33s is a compact, powerful, full featured and affordable system. It provides a detailed flag information never before seen on a 3-part analyzer. The system provides information useful for diagnosis including WBC flag, RBC flag and PLT flag.
  • COMPACT DESIGN
  • COST EFFICIENT
  • Higher throughput at 70 tests per hour
  • MICRO SAMPLE VOLUME
  • Micro sample volume at 9.0uL for whole blood mode with capillary whole blood samples supported, perfect for pediatric samples
  • BETTER CONNECTIVITY
Next generation hematology analyzer.
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms