Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Potassium iodide ≥99.0% AR ACS, Rankem®

Supplier: RANKEM

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


P0461 P0462 P0463
RANKP0461EA 3277.89 INR InStock
RANKP0461 RANKP0462 RANKP0463
Potassium iodide ≥99.0% AR ACS, Rankem®
Potassium iodide
General lab reagent for routine laboratory use.

  • Suitable for routine laboratory usage and classical analysis
  • Available in various pack size

Description: Colourless crystals or white crystalline powder.


Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330 °C (1013 hPa)
Melting Pt: 681 °C
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727
Order Now
Specification Test Results
Guarantee Analysis .
Assay Min. 99.00 %
pH (5% Aqueous Solution) 6.00 --> 9.00
Maximum Limits Of Impurities .
Iodate Passes test
Nitrogen Compounds ≤0.003%
Sulphate (SO4) ≤0.01%
Arsenic (As) ≤ 0.0001%
Barium (Ba) ≤0.002%
Calcium(Ca) ≤0.005%
Heavy Metal (As Pb) ≤0.001%
Iron (Fe) ≤0.002%
Loss On Drying (105 °C) Max. 0.50 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms