Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Tetrahydrofuran ≥99.7% AR ACS, Analytical reagent (AR), Rankem®

Supplier: RANKEM

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

T0031 T0032 T0033
RANKT0031EA 2152.5 INR InStock
RANKT0031 RANKT0032 RANKT0033
Tetrahydrofuran ≥99.7% AR ACS, Analytical reagent (AR), Rankem®
Tetrahydrofuran


Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108,5 °C
Density: 0,888 g/cm³ (20 °C)
Flash Pt: −21,5 °C
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
Merck Index: 12,09351
Order Now
Specification Test Results
Color (APHA) ≤ 10
Guarantee Analysis .
Assay (GC) (C4H8O) ≥ 99.70 %
Bht (Stabilizer) About 250 ppm
Weight Per ml (at 20 °C) 0.887 - 0.889 g
Identity (IR) Conforms
Maximum Limits Of Impurities .
Residue After Evaporation ≤ 0.001 %
Titrable Acid ≤ 0.0002 meq/g
Titrable Base ≤ 0.0002 meq/g
Water ( By KF) ≤ 0.03 %
Peroxide (As H2O2) ≤ 0.005 %
Copper(Cu) ≤ 0.0001 %
Iron (Fe) ≤ 0.0001 %
Lead ( Pb) ≤ 0.0001 %

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms