Mercury(II) chloride ≥97% SG, Laboratory Reagent, Rankem®
Supplier: RANKEM (A AVANTOR BRAND)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Mercury dichloride
, Mercuric chloride
, Mercury Chloride
RANKM0095EA
0
INR
RANKM0095
Mercury(II) chloride ≥97% SG, Laboratory Reagent, Rankem®
Mercury (II) chloride
General lab reagent for routine laboratory use and classical analysis.
- Suitable for routine laboratory usage and classical analysis
Description: White crystals/powder/granules.
Formula:
HgCl₂ MW: 271,5 g/mol Boiling Pt: 302 °C (1013 hPa) Melting Pt: 273 °C Density: 5,44 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011041 CAS Number: 7487-94-7 EINECS: 231-299-8 UN: 1624 ADR: 6.1,II Merck Index: 13,05901 |
Specification Test Results
Guarantee Analysis | . |
Assay ( Complexometric) | Min. 97.00 % |
Maximum Limits Of Impurities | . |
Residue After Reduction | Max. 0.0500 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...