Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® SAS Super 100 and 180 Air Samplers
Supplier: VWR Collection
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
PBIB90717EA 226595.93 INR InStock
PBIB90717 PBIB90716
VWR® SAS Super 100 and 180 Air Samplers
Samplers Air Samplers
SAS Super 100 and SAS Super 180 air samplers provide 100 or 180 litres per minute calibrated flow with real-time electronic control of actual air flow, according to EN 17025 specifications.
  • Delayed start option
  • Active status confirmed by extra-bright warning light
  • Optional IR remote control
  • Microbial validation
  • Software is available for sampling data input
Programmable intervals between samplings enable users to extend each sampling period from a few minutes to several hours, without operator’s supervision. Autoclavable sampler heads are available in chrome-plated aluminium, stainless steel, or plastic.
Certifications: EN 17025, RS232 serial port for data transfer to PC.
Ordering information: For a complete system, please order the required sampler, a battery charger, and the appropriate head.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms