Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Perchloric acid solution 0,1 N, Analytical reagent (AR), Rankem®

Supplier: RANKEM (A AVANTOR BRAND)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Warning

RANKP0049EA 2105.4 INR InStock
RANKP0049
Perchloric acid solution 0,1 N, Analytical reagent (AR), Rankem®
Perchloric acid
High purity analytical reagent.

  • Ready to use solution for 0,1 N

Description: A clear, colourless liquid with a pungent odour.


Formula: HClO₄
MW: 100,46 g/mol
Boiling Pt: 39 °C (75 hPa)
Melting Pt: -101 °C
Storage Temperature: ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 3264
ADR: 8,III
Order Now
Specification Test Results
Normality 0.095 - 0.105 N
Water ≤ 0.02 %

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms