Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Dichloromethane ≥99.50%, Analytical reagent (AR), Rankem®

Supplier: RANKEM

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Warning

Stabilization: Stabilised with amylene

D0071 D0072 D0073
RANKD0071EA 1143.45 INR InStock
RANKD0071 RANKD0072 RANKD0073
Dichloromethane ≥99.50%, Analytical reagent (AR), Rankem®
Dichloromethane
High purity analytical reagent that meets or exceeds the ACS reagent Specifications.

  • Batch-to-Batch consistency
  • Tested for heavy metals
  • Suitable for analytical testing
  • Available in various pack size

A colourless liquid free from matter in suspension having a sweet penetrating ether- like odour.

Soluble in ethyl alcohol and ethyl ether in all proportions.


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
Order Now
Specification Test Results
BIS Specification
Purity ≥ 99.50%
Relative Density at 27/27 °C 1.314 - 1.319
Residue On Evaporation ≤ 0.001%
Moisture ≤ 0.02%
Acidity (as HCl) ≤ 0.0003%
Free Chlorine Passes test
Aldehydes Passes test
Iron (Fe) Max. 0.04 ppm
Heavy Metals (As Pb) ≤ 1.00 ppm
Resistance to potassium permanganate Passes test
Nessler’s reagent Passes test
Resistance to sulphuric acid Passes test
Total impurities due to methyl chloride and chloroform ≤ 0.10%
AR, ACS Specifications
Weight Per ml (at 20 °C) 1.323 - 1.325 g
Identity (IR) Conforms
Colour ≤ 10 APHA
Amylene (As Stabiliser) 30 - 150 ppm
Free Halogens Passes test
Other Chlorinated Organic Impurities ≤ 0.10 %
Copper (Cu) ≤ 0.0001%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms