Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Florisil® 60-100 mesh for chromatography

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: TalcFlorisilMagnesium silicateMagnesium silicate, activated

15025HSEA 0 INR InStock
15025HS
Florisil® 60-100 mesh for chromatography
Magnesium silicate


Formula: MgO₃Si
MW: 100,38 g/mol
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1
Order Now

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms