Acrylogel 3 solution 40%, Electran for electrophoresis, VWR®
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
443733REA 0 INR
VWRC443733R
Acrylogel 3 solution 40%, Electran for electrophoresis, VWR®
A convenient, ready to use 40% w/v (40%T) solution of acrylamide Electran and NN'-methylenebisacrylamide Electran 3% (C±0,2%) in deionised water. Final ratio 29,1:0,9.