Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Polyvinylpyrrolidone, M.W. ∼ 360,000, Electran for molecular biology (K90)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: PolyvidonePolyvinylpyrrolidonPVP

436032CEA 0 INR InStock
436032C
Polyvinylpyrrolidone, M.W. ∼ 360,000, Electran for molecular biology (K90)
Polyvinylpyrrolidone


Formula: (C₆H₉NO)n
MW: 360000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
UN: 0000
Order Now
Specification Test Results
N (Nitrogen) 11.50 - 12.80 %
Identity (IR) Passes test
DNases Not detected
RNases Not detected
Proteases Not detected
Heavy metals (as Pb) Max. 0.001 %
Loss on drying (105°C) Max. 5.00 %
pH (20°C; 1 %) 5.00 - 8.00

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms