Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Conductivity standard solutions, Orion™
Supplier: Thermo Fisher Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Orion™
ORIO011005EA 0 INR InStock
ORIO011005 663-0024 663-0023 663-0044
Conductivity standard solutions, Orion™
Standards Conductivity Standards
Conductivity standard temperature tables are programmed into Thermo Scientific™ Orion™ meter software for use with the automatic calibration feature.
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms