Nitrochromic reagent Reag. Ph. Eur. 1059100
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
87881.180EA
9985
INR
87881.180
Nitrochromic reagent Reag. Ph. Eur. 1059100
Nitrochromic reagent
The solution bottle should be open for the minimum time required to dispense the solution.
After use, the bottle should be tightly recapped and stored under normal laboratory conditions.
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Storage Temperature:
Ambient |
MDL Number:
MFCD00011367 EINECS: 000-000-0 UN: 3289 ADR: 6.1 (8),I |
Specification Test Results
Starting materials | potassium dichromate R / nitric acid R |
Performance test | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...