Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Microplate lids, Robolids
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
732-5507EA 0 INR InStock
CORN3090
Microplate lids, Robolids
Made from PS. Combines the sealing ability of a storage mat with the rigidity of a plastic lid.
  • Designed for repeated application and removal by automation and to prevent short-term evaporation
  • Silicone sealing plugs for organic solvent resistance and low extractables
  • Can be used manually or with automation
  • Non sterile
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms