Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours. Complete with integrated air valve for pressure equalisation. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
- Especially recommended for HPLC systems
- Chemically resistant against aggressive organic solvents
Renew air valve twice a year for maximum safety.
SafetyCaps with shut-off: During repairs on HPLC pumps, the shut-off on the SafetyCaps can be closed to keep capillaries and inlet filters in the solvent. The frit does not come into contact with the ambient air. The pump can be quickly and easily flushed during purging after repairs. The SafetyCaps with shut-off also prevent air pockets in the tube when the supply bottle is being changed and guarantees that the analysis continues without a problem.
SafetyCaps combined: The SafetyCap combi, and its all-around connector, provide a stable baseline and reliable reproducibility for RI detection. For fast and simple solvent degassing with helium.
SafetyCaps for NS 29/32 ground neck bottles: The locknut allows the stopper to be easily loosened and removed, even after long usage. For bottles with NS 29/32 mm standard ground joints.
Delivery information: All SafetyCaps are delivered with an air valve and corresponding fittings.