Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Tough-Tags® label rolls
Supplier: Diversified Biotech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Tough-Tags®
817-2133EA 0 INR InStock
817-2129 817-2128 817-2116 DIVETTSO-1000 817-2115 817-2114 817-2135 817-2113 817-2134 817-2112 817-2133 817-2111 817-2132 817-0052 DIVETTLO-1000 DIVETTGP-1000 817-2130
Tough-Tags® label rolls
Labels
These Tough-Tags® polyester label rolls are pre-cut, peel-off labels which accept many marking instruments. They are perfectly sized to fit microcentrifuge tubes and other containers in the laboratory.
  • Temperature resistant from −40 to +121 °C
  • Chemically inert, resist most organic solvents and caustic agents
  • Adhere to most plastics, glass and metals without cracking, peeling or degrading
  • Can be written on with pen, pencil or permanent marker
Tough-Tags® resist boiling water baths, autoclaving and moderate freezer temperatures. Tubes labeled with Tough-Tags® easily slide in and out of centrifuge rotors without binding.
Order Now
ORDER
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms