Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Disposable half masks against gases, vapours and particulates, Freedom
Supplier: Honeywell Safety Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Freedom
111-3531EA 0 INR InStock
DALL1001598 DALL1001591
Disposable half masks against gases, vapours and particulates, Freedom
Half face masks with integrated filters for occasional or frequent exposures to gases, vapours and particulates.
  • Quick and easy adjustment
  • Excellent fit and comfort due to a soft seal flange
  • Disposable - no maintenance, better hygiene and no filter handling
  • Expiration valve for easier breathing
Weight: 265 g
Certifications: EN 405, EN 149: 2001
E: Acidic gases, e.g. sulphur dioxide, hydrogen chloride
Caution: *One size fits all
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms