Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Capillary Tips for Gel Loading
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
732-0509EA 4589 INR InStock
732-0509 732-0508
VWR®, Capillary Tips for Gel Loading
Pipette Tips
Non bevelled capillary section, non graduated, compatibility type H.
  • Thin capillary fits between gel plates
  • Accurately fills the wells between the teeth of the gel comb
  • Wide sealing areas to also fit older pipette barrels
  • Virgin resins and sustainable product design
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms