Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Test Tube and Drying Racks
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
211-0222EA 888 INR InStock
211-0222
VWR®, Test Tube and Drying Racks
Racks Tube Racks
PP, reinforced.
  • Heat resistant up to 135 °C, autoclavable
  • Do not float in a water bath
  • Excellent chemical resistance
Secure hold for test tubes, sample tubes and reaction vessels. Can be used as drying racks for test tubes, electrophoresis and chromatography plates.
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms