Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Microcentrifuge tubes, 1,7 ml, SuperClear boil-proof
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
211-0032EA 2520 INR InStock
VWRI211-0032
Microcentrifuge tubes, 1,7 ml, SuperClear boil-proof
Ultra-clear PP, graduated, autoclavable and freezable, without cap.
  • Can withstand centrifugation to 20000×g
  • Fits all standard and high capacity rotors
  • Sustainable design based on virgin resins
  • Every lot is tested and guaranteed DNase- and RNase-free
Packaging: In resealable bags.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms