Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Optimizer PCR Workstation™
Supplier: CBS Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
730-0776EA 0 INR InStock
135-2045 730-0790 135-2044 135-2058 135-2046 135-2059 730-0776 730-0764 135-2061 135-2060 135-2052 730-0772 135-2051 135-2054 730-0792 730-0780 135-2053 732-2563 730-0788
Optimizer PCR Workstation™
Fume Hoods and Enclosures PCR Workstation Enclosures
Acid-resistant black formica or durable stainless steel work surface, tempered safety glass fascia.
  • Access doors close to prevent contamination, and slide into a storage compartment in the base during experimental procedures
  • Hinged glass fascia can be easily lifted out of the way for placement and removal of a thermal cycler, or other large equipment
  • Dual fluorescent lamps provide work surface with excellent visibility
  • Dual UV bulb option delivers twice the intensity of UV light and helps to irradiate areas that might otherwise be inaccessible, particularly recommended when the Workstation is being used to decontaminate apparatus and reagents
  • Safety interlock automatically shuts off UV light when hood is opened
The Optimizer PCR Workstation™ is designed to protect against contamination in sensitive PCR amplification reactions in two ways. Firstly, UV irradiation of the working area prior to use blocks replication of contaminating DNA sequences by causing adjacent pyrimidines to undergo dimerisation. Secondly, it protects against cross or airborne contamination by limiting exposure of the experimental set-up to the open laboratory environment. Available in three sizes, each with plenty of interior space for PCR reaction supplies and thermal cycler. A single electrical outlet and two fluorescent lamps are mounted in the stainless steel ceiling. Each workstation is equipped with a 12 hour countdown timer with time-hold position.
Order Now
ORDER
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms