Trinocular microscope, IT 400
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
630-0792EA 0 INR
VWRI630-0792 VWRI630-0766
Trinocular microscope, IT 400
This inverted microscope is suitable for tissue culture and live cell applications.
  • Working distance of 50 mm
  • Stage for use with 96-well culture microplates and Petri dishes
  • Condenser can be swung out for observing larger vessels (roller bottles or cell factories)
Designed for use in research facilities, universities, medical establishments, and agricultural research institutions.
Delivery information: Supplied with protective dust cover, spare halogen bulb and fuse.