Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Filters, Aquatron®
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
171-0073EA 0 INR InStock
STUAAFH
Filters, Aquatron®
Removes coarse particles, e.g. rust and silt from the water supply using a disposable 10 micron polypropylene filter. May be connected directly to the Aquatron® water still, or linked in series with deioniser 171-1211 using coupling 171-1216.
Delivery information: Supplied with tubing and a tap adapter which fits most taps. Disposable cartridges 171-1214 (illustrated) must be ordered separately.
Caution: The filter unit can be wall mounted using bracket 171-1215
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms