Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Water still, Distinction D4000
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
stuart®
171-1205EA 0 INR InStock
171-1205
Water still, Distinction D4000
Water Purification Systems Stills
The all glass construction and silica sheathed heating element ensure exceptionally high quality pyrogen-free distillate (although distillate should be tested). Fitted with two independent thermostats to prevent overheating in the event of cooling water supply failure; one is mounted in the condenser vent tube to quickly detect water supply failure and the second is mounted inside the boiler. Either will turn off the heater if the water supply becomes too Low. The second thermostat also acts as a backup to the first for extra security. As there is no metal in contact with the water, the Distinction D4000 eliminates the risk of metal ions Leeching out into the water causing contamination to the distillate. The unit is also fitted with a Level control device that will automatically turn off the heater when the reservoir is full. This allows the still to run unattended without risk of the reservoir overflowing.
  • Rapid, reliable, convenient and economical
  • Support stand allows either bench or wall installation
  • Built-in PTFE drain stopcock and acid funnel allow easy cleaning without dismantling the glassware
  • Unique condenser design ensures that the droplets remain in contact with the cooling coil thereby producing cold distilled water
pH, conductivity and resistivity are affected by temperature and the presence of absorbed carbon dioxide. Specifications are based on tests conducted at 20 °C with CO₂-free water.
Care is needed to obtain pyrogen-free water and the distillate must be tested before use.
Water still heating elements are excluded from the 3 year warranty and are covered for 1000 hours use.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms