Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Cryostorage boxes
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
211-0373EA 0 INR InStock
VWRU16466-016 VWRU16466-014 VWRU16466-024 VWRU16466-022
Cryostorage boxes
Boxes are made from polypropylene and come with an alphanumerically labelled grid for identifying contents.
The 50- and 100-place freezer boxes come with attached hinged lid and snap lock closure for versatility. They are designed to hold 1,5 and 2 ml tubes and some 1,2 ml cryo vials. Indented lids can be labelled.
The 81-place box is designed to fit into existing freezer storage racks, holds 1,5 or 2,0 ml microcentrifuge tubes, and includes an attached clear lid. Boxes are autoclavable. Freezable to –90 °C.
Ordering information: Assorted colours packages include one each in blue, green, orange, red, and yellow.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms