Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Testing Sieves, All Stainless Steel
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
510-1584EA 0 INR InStock
510-1584 510-1586 510-1585 510-1500 510-1588 510-1467 510-1587 510-1502 510-1469 510-1468 510-1501 510-1589 510-1515 510-1514 510-1517 510-1516 510-1519 510-1518 510-1485 510-1484 510-1520 510-1487 510-1486 510-1522 510-1489 510-1488 510-1521 510-1524 510-1523 510-1481 510-1480 510-1483 510-1482 510-1504 510-1506 510-1505 510-1508 510-1507 510-1509 510-1474 510-1473 510-1476 510-1475 510-1511 510-1478 510-1477 510-1513 510-1479 510-1512 510-1470 510-1590 510-1472 510-1537 510-1536 510-1539 510-1538 510-1540 510-1542 510-1541 510-1544 510-1543 510-1546 510-1545 510-1526 510-1525 510-1528 510-1527 510-1529 510-1496 510-1495 510-1531 510-1498 510-1497 510-1530 510-1533 510-1532 510-1499 510-1535 510-1534 510-1490 510-1492 510-1491 510-1494 510-1493 510-1559 510-1558 510-1562 510-1561 510-1563 510-1560 510-1548 510-1547 510-1549 510-1551 510-1550 510-1553 510-1552 510-1555 510-1554 510-1557 510-1556
VWR® Testing Sieves, All Stainless Steel
Sieves Test Sieves
Brass wire cloth and frame for higher corrosion resistance than sieves containing brass wire cloth or frames. High durability and wear resistance. Less likelihood of metallic sample contamination. Full coarse and fine mesh range down to 20 µm (0.0008"). Sturdy construction with fillet solder bead between the frame and wire cloth to prevent particles from being trapped. Diameter: 20,3 cm (8"). Choose full height with 5,08cm (2") depth to wire, or half height with 2,54 cm (1") depth to wire.
Inspected with an optical measurement system traceable to the National Institute of Standards and Technology. Each sieve bears a traceable serial number and comes with certificate of compliance to applicable ASTM E-11 and ISO specifications.
CENTERLINE® measurement (47730-590) is a NIST traceable service that checks calibration and gives statistical measurement information on each sieve.
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms