Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Bottle-top dispensers, QuikSip™
Supplier: Brand
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
QuikSip™
612-5638EA 0 INR InStock
612-5638
Bottle-top dispensers, QuikSip™
Bottle Top Dispensers
Variable volume, partly autoclavable.
  • Safe removal of supernatants (up to maximum 25 ml per plunger stroke)
  • Works without vacuum pump
  • Fingertip vacuum control using the cell-culture™ unit
  • Works as single channel or 8-channel aspiratior (manifold optional)
  • For use with disposable pipette tips, micropipettes and glass pasteur pipettes
The QuikSip™ bottletop dispenser is designed for safe and fast aspiration of common laboratory liquids used in biology, food chemistry and medicine, example biological solutions, nutrient media, polar solvents, aqueous solutions. The innovative design permits quick disassembly. Adapter and suction tube of the cell-culture™ are autoclavable at 121 °C (2 bar), the dispensing cartrige and pump unit are not autoclavable.
Delivery information: Supplied with 1 cell-culture™-unit including suction tube and three adapters, operating manual, spare dispensing cartridge and two PP adapters (GL 45/32 and GL 45/S 40).
Order Now
ORDER
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms